Assay Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF295102.1 | - | 1 - 2 | 64 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF295102.1 transcripts.
Set Membership: |
|

Scamp4
Genomic Map