Assay Details
| Gene Symbol: Tekt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011902.2 | NP_036032.2 | 3 - 4 | 402 | 87 | |
| GenBank mRNA | AB027138.1 | - | 3 - 4 | 402 | 87 | |
| BC008140.1 | - | 3 - 4 | 3582971 | 87 | ||
| BQ712964.1 | - | 3 - 4 | 416 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tekt2
Genomic Map