Assay Details
| Gene Symbol: Dlgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158868.1 | - | 7 - 8 | 2527 | 66 | |
| AY243848.2 | - | 5 - 6 | 1907 | 66 | ||
| BC057615.1 | - | 7 - 8 | 2136 | 6401651 | 66 | |
| BC058433.1 | - | 5 - 6 | 1334 | 66 | ||
| BC098311.1 | - | 2669 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap3
Genomic Map