Assay Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164316.1 | NP_001157788.1 | 2 - 3 | 1642 | 140 | |
| NM_183310.2 | NP_899133.2 | 2 - 3 | 1694 | 140 | ||
| GenBank mRNA | AK173231.1 | - | 1 - 2 | 387 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccser1
Genomic Map