Assay Details
| Gene Symbol: Sema4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF038652.1 | - | 9 - 10 | 1229 | 105 | |
| AK035993.1 | - | 10 - 11 | 1502 | 105 | ||
| AK047067.1 | - | 10 - 11 | 1486 | 105 | ||
| AK143196.1 | - | 10 - 11 | 1514 | 105 | ||
| BC057048.1 | - | 9 - 10 | 1378 | 105 | ||
| BC090625.1 | - | 10 - 11 | 1470 | 105 | ||
| BC094567.1 | - | 10 - 11 | 1497 | 6834138 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sema4f
Genomic Map