Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 2 - 3 | 281 | 60 | |
| GenBank mRNA | AK041141.1 | - | 2 - 3 | 254 | 60 | |
| AK143510.1 | - | 2 - 3 | 245 | 60 | ||
| AK162776.1 | - | 2 - 3 | 245 | 60 | ||
| BB636410.1 | - | 2 - 3 | 254 | 60 | ||
| BC015245.1 | - | 2 - 3 | 218 | 4242193 | 60 | |
| CJ059053.1 | - | 2 - 3 | 282 | 60 | ||
| L03292.2 | - | 2 - 3 | 253 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041141.1, AK143510.1, BC015245.1, L03292.2, NM_009320.4 transcripts.
Set Membership: |
|

Slc6a6
Genomic Map