Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 7 - 8 | 1159 | 89 | |
| GenBank mRNA | AK143510.1 | - | 7 - 8 | 1123 | 89 | |
| BC015245.1 | - | 7 - 8 | 1096 | 4242193 | 89 | |
| L03292.2 | - | 7 - 8 | 1131 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a6
Genomic Map