Assay Details
| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198884.1 | NP_942585.1 | 18 - 19 | 2723 | 75 | |
| GenBank mRNA | AB114826.1 | - | 18 - 19 | 2723 | 75 | |
| AK082874.1 | - | 4 - 5 | 442 | 76 | ||
| AK143898.1 | - | 18 - 19 | 2639 | 75 | ||
| BC139798.1 | - | 18 - 19 | 2737 | 40060840 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

B4galnt3
Genomic Map