Assay Details
| Gene Symbol: Ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286405.1 | NP_001273334.1 | 2 - 3 | 255 | 73 | |
| GenBank mRNA | AY179430.1 | - | 2 - 3 | 245 | 73 | |
| BC094911.1 | - | 2 - 3 | 259 | 73 | ||
More Information
Set Membership: |
|

Ghrl
Genomic Map