Assay Details
| Gene Symbol: Zbtb24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153398.3 | NP_700447.2 | 6 - 7 | 1567 | 57 | |
| NR_102360.1 | 6 - 7 | 1562 | 57 | |||
| GenBank mRNA | AK054097.1 | - | 6 - 7 | 1523 | 57 | |
| AK146986.1 | - | 6 - 7 | 1543 | 57 | ||
| AK154467.1 | - | 6 - 7 | 1538 | 57 | ||
| AY210402.1 | - | 6 - 7 | 1541 | 57 | ||
| BC050933.1 | - | 6 - 7 | 1529 | 6811382 | 57 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089285 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074197 | BC055367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074197 | AY210402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074197 | BC038139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074197 | BC050933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074197 | AK154467.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167355 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111776 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147861 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074197 | BC055367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074197 | AY210402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074197 | BC038139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074197 | BC050933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074197 | AK154467.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P302108 | BC055367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P302108 | AY210402.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P302108 | BC050933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P302108 | AK154467.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P302108 | AK169260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174542 | BC055367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174542 | AY210402.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174542 | BC038139.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174542 | BC050933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174542 | AK154467.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27089283 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111776 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07038554 | - | - |
| Whole Mouse Genome G4122A | A_51_P174542 | BC055367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174542 | AY210402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174542 | BC038139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174542 | BC050933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174542 | AK154467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P302108 | BC055367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P302108 | AY210402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P302108 | BC050933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P302108 | AK154467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P302108 | AK169260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209451 | AF263010.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139802 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130063 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147861 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08963675 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08779365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08779364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08641895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08641898 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08963672 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08963662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089284 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27089285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08616629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08209332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08461232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08461228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08855324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08618371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08617205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089294 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10362764 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362766 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362761 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362765 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362762 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362763 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438218_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442567_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362758 | - | - |
| Mouse Genome 430 2.0 Array | 1442567_at | - | - |
| Mouse Genome 430 2.0 Array | 1438218_at | - | - |
| Mouse Genome 430 2.0 Array | 1446628_at | - | - |
| Mouse Genome 430 2.0 Array | 1438218_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446628_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442567_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 268294_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 268294_PM_at | - | - |
| Mouse Expression Set 430 | 1438218_at | - | - |
| Mouse Expression Set 430 | 1442567_at | - | - |
| Mouse Expression Set 430 | 1442567_PM_at | - | - |
| Mouse Expression Set 430 | 1438218_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438218_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438218_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442567_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442567_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4570504 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570504 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234461 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234461 | - | - |
| mouse-6 v1.1 (Illumina) | scl38986.7.1_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570504 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38986.7.1_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570504 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234461 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234461 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38986.7.1_55-S | - | - |

Zbtb24
Genomic Map