Assay Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007482.3 | NP_031508.1 | 5 - 6 | 656 | 108 | |
| GenBank mRNA | AK149471.1 | - | 5 - 6 | 639 | 108 | |
| AK171417.1 | - | 4 - 5 | 490 | 108 | ||
| BC013341.1 | - | 5 - 6 | 633 | 4163626 | 108 | |
| BC050005.1 | - | 5 - 6 | 670 | 3990246 | 108 | |
| CT010173.1 | - | 5 - 6 | 554 | 108 | ||
| U51805.1 | - | 5 - 6 | 695 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arg1
Genomic Map