Assay Details
| Gene Symbol: 4930587E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016361.1 | - | 2 - 3 | 122 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | contactin 3 | Cntn3 | Pang | Anti-sense | Chr. 6 - 102163304 - 102564620 on NCBI Build GRCm38 | |
| Downstream | microRNA 6373 | Mir6373 |
Gm27870, mmu-mir-6373 |
524.4kb | Sense | Chr. 6 - 102794384 - 102794485 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930587E11Rik
Genomic Map