Assay Details
| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 7 - 8 | 1026 | 70 | |
| NM_001080747.2 | NP_001074216.1 | 7 - 8 | 1026 | 70 | ||
| NM_001080748.2 | NP_001074217.1 | 7 - 8 | 1026 | 70 | ||
| NM_001080749.2 | NP_001074218.1 | 7 - 8 | 1026 | 70 | ||
| NM_010365.4 | NP_034495.2 | 7 - 8 | 1026 | 70 | ||
| GenBank mRNA | AF017085.1 | - | 8 - 9 | 730 | 70 | |
| AF043219.1 | - | 6 - 7 | 647 | 70 | ||
| AF043220.1 | - | 6 - 7 | 647 | 70 | ||
| AK147201.1 | - | 7 - 8 | 984 | 70 | ||
| AY030290.1 | - | 7 - 8 | 719 | 70 | ||
| AY030291.1 | - | 7 - 8 | 719 | 70 | ||
| AY030292.1 | - | 7 - 8 | 719 | 70 | ||
| AY030293.1 | - | 7 - 8 | 719 | 70 | ||
| BC047387.1 | - | 7 - 8 | 674 | 70 | ||
| BC053044.1 | - | 7 - 8 | 995 | 5704935 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2i
Genomic Map