Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 8 - 9 | 959 | 146 | |
| GenBank mRNA | AF086905.1 | - | 7 - 8 | 920 | 146 | |
| AK044913.1 | - | 8 - 9 | 960 | 146 | ||
| AK170235.1 | - | 7 - 8 | 903 | 146 | ||
| BC056617.1 | - | 8 - 9 | 983 | 4977960 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek2
Genomic Map