Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 9 - 10 | 1059 | 77 | |
| GenBank mRNA | AF086905.1 | - | 8 - 9 | 1020 | 77 | |
| AK044913.1 | - | 9 - 10 | 1060 | 77 | ||
| AK170235.1 | - | 8 - 9 | 1003 | 77 | ||
| BC056617.1 | - | 9 - 10 | 1083 | 4977960 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek2
Genomic Map