Assay Details
| Gene Symbol: Gm2861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080443.1 | - | 99 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33609 |
Gm33609 | -45.7kb | Sense | Chr. 5 - 97131774 - 97149849 on NCBI Build GRCm38 | |
| Downstream |
N(alpha)-acetyltransferase 11, NatA catalytic subunit |
Naa11 |
Ard1b, Ard2, C80008 |
174.3kb | Anti-sense | Chr. 5 - 97382209 - 97392330 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | AK044765.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | AK047184.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | AK049753.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | BC034054.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | BC049954.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549834 | NM_146025.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05663767 | - | - |
| Whole Mouse Genome G4122A | A_52_P787779 | AK080443.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103193 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05663767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04569739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04642881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04554415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04112452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04569734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04569728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04649168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04641007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04499360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04569729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04499351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04950927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04138365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04771015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04628309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04266573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23911227 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 540177 | - | - |

Gm2861
Genomic Map