| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900013266.mm.1 |
NM_001163495.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013266.mm.1 |
NM_001163495.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013266.mm.1 |
NM_027667.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013266.mm.1 |
NM_027667.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013266.mm.1 |
TC1900001510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900013267.mm.1 |
NM_001163495.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013267.mm.1 |
NM_001163495.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013267.mm.1 |
NM_027667.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013267.mm.1 |
NM_027667.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900013267.mm.1 |
TC1900001510.mm.1 |
MTA TC |