Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK150218.1 | - | 1 - 2 | 481 | 151 | |
| BC019498.1 | - | 1 - 2 | 440 | 4460879 | 151 | |
| BC057658.1 | - | 1 - 2 | 121 | 3153717 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150218.1, BC019498.1, BC057658.1 transcripts.
Set Membership: |
|

Fam216a
Genomic Map