Assay Details
| Gene Symbol: Sis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081137.1 | NP_001074606.1 | 12 - 13 | 1487 | 82 | |
| GenBank mRNA | EU937530.1 | - | 13 - 14 | 1550 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sis
Genomic Map