Assay Details
| Gene Symbol: Adam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272057.1 | NP_001258986.1 | 4 - 5 | 643 | 104 | |
| NM_001272058.1 | NP_001258987.1 | 4 - 5 | 643 | 104 | ||
| NM_001272059.1 | NP_001258988.1 | 4 - 5 | 643 | 104 | ||
| NM_007401.3 | NP_031427.2 | 4 - 5 | 643 | 104 | ||
| GenBank mRNA | AK016574.1 | - | 4 - 5 | 299 | 104 | |
| AK029519.1 | - | 4 - 5 | 333 | 104 | ||
| AK161262.1 | - | 4 - 5 | 643 | 104 | ||
| AK161417.1 | - | 4 - 5 | 313 | 104 | ||
| BC094237.1 | - | 4 - 5 | 340 | 30604517 | 104 | |
| U22059.1 | - | 4 - 5 | 307 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam5
Genomic Map