Assay Details
| Gene Symbol: Whsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081102.2 | NP_001074571.2 | 9 - 10 | 2045 | 80 | |
| NM_175231.2 | NP_780440.2 | 9 - 10 | 2042 | 80 | ||
| GenBank mRNA | AK078622.1 | - | 6 - 7 | 680 | 80 | |
| AK129287.1 | - | 6 - 7 | 980 | 80 | ||
| BC046473.1 | - | 4 - 5 | 471 | 80 | ||
| BC079668.1 | - | 9 - 10 | 2068 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078622.1 transcripts.
Set Membership: |
|

Whsc1
Genomic Map