Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 21 - 22 | 2027 | 140 | |
| GenBank mRNA | AB008516.1 | - | 20 - 21 | 1904 | 140 | |
| AK053765.1 | - | 21 - 22 | 2131 | 140 | ||
| AK084201.1 | - | 11 - 12 | 1062 | 140 | ||
| AK088273.1 | - | 20 - 21 | 1974 | 140 | ||
| AK165850.1 | - | 10 - 11 | 935 | 140 | ||
| AK166120.1 | - | 11 - 12 | 967 | 140 | ||
| AK166164.1 | - | 12 - 13 | 1116 | 140 | ||
| AK220495.1 | - | 21 - 22 | 2069 | 140 | ||
| BC019173.1 | - | 21 - 22 | 1996 | 140 | ||
| BC037654.1 | - | 20 - 21 | 1927 | 140 | ||
| BC057207.1 | - | 20 - 21 | 1931 | 140 | ||
| BC145489.1 | - | 16 - 17 | 1547 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map