Assay Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173189.2 | NP_775281.2 | 9 - 10 | 2317 | 79 | |
| GenBank mRNA | AK042026.1 | - | 8 - 9 | 2211 | 79 | |
| AK148021.1 | - | 9 - 10 | 1915 | 79 | ||
| AY070216.1 | - | 9 - 10 | 1897 | 79 | ||
| BC056924.1 | - | 9 - 10 | 2317 | 5697978 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcph1
Genomic Map