Assay Details
| Gene Symbol: Mef2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133665.3 | NP_598426.1 | 5 - 6 | 1015 | 101 | |
| GenBank mRNA | AK141270.1 | - | 5 - 6 | 995 | 101 | |
| AK155778.1 | - | 5 - 6 | 1016 | 101 | ||
| BC011070.1 | - | 5 - 6 | 983 | 3497896 | 101 | |
| S68893.1 | - | 5 - 6 | 837 | 101 | ||
| S68895.1 | - | 5 - 6 | 834 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2d
Genomic Map