| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1431228_PM_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1431228_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1431228_PM_s_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900014115.mm.1 |
NR_015516.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014115.mm.1 |
NR_027950.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014115.mm.1 |
NR_102721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014115.mm.1 |
TC0900001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900014114.mm.1 |
NR_015516.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014114.mm.1 |
NR_027950.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014114.mm.1 |
TC0900001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900014116.mm.1 |
NR_015516.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014116.mm.1 |
NR_027950.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014116.mm.1 |
NR_102721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900014116.mm.1 |
TC0900001679.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1431228_PM_s_at |
- |
- |
| Mouse Expression Set 430 |
1431228_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1431228_PM_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1431228_s_at |
- |
- |