Assay Details
| Gene Symbol: Pea15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY487415.1 | - | 348 | 67 | ||
| BC132661.1 | - | 445 | 40130938 | 67 | ||
| BC132663.1 | - | 445 | 40130940 | 67 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | NM_001010832.1 | - | 348 | Chr. - - [-] on Build | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059139 | BC132661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059139 | BC132663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2059139 | NR_027806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091012 | AY487415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091012 | BC132661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091012 | BC132663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091012 | NR_027806.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04551267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04936211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04465849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04800608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04626408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04842469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04839620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04200473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04602927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04626411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04865367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04936210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04865270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04248800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04790965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04551259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04551270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04758419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04989706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04200470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04758423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04865267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04505828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04949273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04839624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04626407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23822139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10530848 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10530847 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500023453.mm.1 | TC0500002652.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023454.mm.1 | TC0500002652.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023452.mm.1 | TC0500002652.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023455.mm.1 | TC0500002652.mm.1 | MTA TC |

Pea15b
Genomic Map