Assay Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111272.1 | NP_001104742.1 | 10 - 11 | 1305 | 91 | |
| NM_025303.3 | NP_079579.2 | 11 - 12 | 1436 | 91 | ||
| GenBank mRNA | AF459099.1 | - | 11 - 12 | 1386 | 91 | |
| AF459100.1 | - | 10 - 11 | 1255 | 91 | ||
| AK015087.1 | - | 4 - 5 | 385 | 91 | ||
| AK031102.1 | - | 11 - 12 | 1363 | 91 | ||
| AK171076.1 | - | 11 - 12 | 1402 | 91 | ||
| BC010300.1 | - | 8 - 9 | 1111 | 91 | ||
| BC025118.1 | - | 10 - 11 | 1260 | 3670067 | 91 | |
More Information
Set Membership: |
|

Stau2
Genomic Map