Assay Details
| Gene Symbol: Dach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038610.2 | NP_001033699.1 | 4 - 5 | 1737 | 75 | |
| NM_007826.3 | NP_031852.1 | 5 - 6 | 1893 | 75 | ||
| GenBank mRNA | AF129510.1 | - | 4 - 5 | 1499 | 75 | |
| AK047409.1 | - | 4 - 5 | 603 | 75 | ||
| AK053594.1 | - | 4 - 5 | 946 | 75 | ||
| AK139060.1 | - | 4 - 5 | 1738 | 75 | ||
| BC052005.1 | - | 3 - 4 | 981 | 75 | ||
| BC059934.1 | - | 4 - 5 | 1515 | 75 | ||
| BC078644.1 | - | 4 - 5 | 1271 | 75 | ||
| BC145440.1 | - | 4 - 5 | 1343 | 75 | ||
More Information
Set Membership: |
|

Dach1
Genomic Map