Assay Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037997.3 | NP_001033086.2 | 13 - 14 | 1849 | 191 | |
| NM_001286415.1 | NP_001273344.1 | 12 - 13 | 1677 | 191 | ||
| NM_008000.2 | NP_032026.2 | 8 - 9 | 1131 | 191 | ||
| GenBank mRNA | AK082799.1 | - | 10 - 11 | 1342 | 191 | |
| AK157828.1 | - | 13 - 14 | 1813 | 191 | ||
| BC051249.1 | - | 5 - 6 | 593 | 191 | ||
| BC058100.1 | - | 12 - 13 | 1677 | 6813507 | 191 | |
| M32054.1 | - | 8 - 9 | 1131 | 191 | ||
| U76762.1 | - | 12 - 13 | 1782 | 191 | ||
More Information
Set Membership: |
|

Fer
Genomic Map