Assay Details
| Gene Symbol: Calb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007586.1 | NP_031612.1 | 5 - 6 | 467 | 92 | |
| GenBank mRNA | AK032171.1 | - | 473 | 92 | ||
| BC017646.1 | - | 5 - 6 | 467 | 4527074 | 92 | |
| X73985.1 | - | 3 - 4 | 156 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calb2
Genomic Map