Assay Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 7 - 8 | 1017 | 75 | |
| NM_016857.2 | NP_058553.2 | 8 - 9 | 1110 | 75 | ||
| GenBank mRNA | AK087601.1 | - | 7 - 8 | 1005 | 75 | |
| AK140450.1 | - | 8 - 9 | 1084 | 75 | ||
| AK162868.1 | - | 8 - 9 | 1097 | 75 | ||
| AK167769.1 | - | 7 - 8 | 1000 | 75 | ||
| AK169928.1 | - | 7 - 8 | 1004 | 75 | ||
| AK173094.1 | - | 6 - 7 | 5597 | 75 | ||
| BC028927.1 | - | 7 - 8 | 993 | 4208803 | 75 | |
| BC053710.1 | - | 7 - 8 | 1004 | 30056570 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc7
Genomic Map