| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100019548.mm.1 |
NM_013770.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019548.mm.1 |
NM_013770.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019548.mm.1 |
TC1100002030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019547.mm.1 |
NM_013770.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019547.mm.1 |
NM_013770.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019547.mm.1 |
TC1100002030.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019543.mm.1 |
NM_013770.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019543.mm.1 |
NM_013770.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019543.mm.1 |
TC1100002030.mm.1 |
MTA TC |