Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 3 - 4 | 318 | 70 | |
| GenBank mRNA | AK134274.1 | - | 3 - 4 | 319 | 70 | |
| AK138442.1 | - | 3 - 4 | 342 | 70 | ||
| AK146469.1 | - | 3 - 4 | 342 | 70 | ||
| BC011321.1 | - | 2 - 3 | 195 | 4007058 | 70 | |
| BC024730.1 | - | 3 - 4 | 318 | 70 | ||
| BC024981.1 | - | 3 - 4 | 318 | 70 | ||
| BC033398.1 | - | 3 - 4 | 318 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC011321.1 transcripts.
Set Membership: |
|

Ddx27
Genomic Map