Assay Details
| Gene Symbol: Dpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010072.3 | NP_034202.1 | 3 - 4 | 325 | 81 | |
| GenBank mRNA | AB004789.1 | - | 3 - 4 | 299 | 81 | |
| AI099209.1 | - | 3 - 4 | 325 | 81 | ||
| AK004834.1 | - | 3 - 4 | 318 | 81 | ||
| AK005268.1 | - | 3 - 4 | 311 | 81 | ||
| AK008548.1 | - | 3 - 4 | 313 | 81 | ||
| BC061151.1 | - | 3 - 4 | 305 | 30307514 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpm1
Genomic Map