Assay Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028131.3 | NP_082407.1 | 10 - 11 | 974 | 102 | |
| GenBank mRNA | AK012097.1 | - | 10 - 11 | 975 | 102 | |
| BC027119.1 | - | 7 - 8 | 1698 | 102 | ||
| BC058243.1 | - | 8 - 9 | 885 | 6309162 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpn
Genomic Map