Assay Details
| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114079.2 | NP_001107551.1 | 3 - 4 | 761 | 65 | |
| GenBank mRNA | BC158030.1 | - | 3 - 4 | 761 | 9007333 | 65 |
| EU669908.1 | - | 3 - 4 | 504 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pabpc1l
Genomic Map