Assay Details
| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 5 - 6 | 849 | 97 | |
| GenBank mRNA | AK076526.1 | - | 1 - 2 | 146 | 97 | |
| AK088016.1 | - | 5 - 6 | 750 | 97 | ||
| AK088729.1 | - | 5 - 6 | 596 | 97 | ||
| AK135521.1 | - | 4 - 5 | 525 | 97 | ||
| AK159689.1 | - | 5 - 6 | 691 | 97 | ||
| AK167682.1 | - | 5 - 6 | 747 | 97 | ||
| BC050906.1 | - | 5 - 6 | 722 | 5721348 | 97 | |
| X15202.1 | - | 4 - 5 | 542 | 97 | ||
| Y00769.1 | - | 5 - 6 | 637 | 97 | ||
More Information
Set Membership: |
|

Itgb1
Genomic Map