Assay Details
| Gene Symbol: Pgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163314.2 | NP_001156786.1 | 18 - 19 | 1829 | 69 | |
| GenBank mRNA | CK392830.1 | - | 5 - 6 | 363 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor metabolic process
GPI anchor biosynthetic process
transport
intracellular protein transport
sensory perception of sound
embryonic pattern specification
anterior/posterior axis specification
protein transport
myo-inositol transport
forebrain regionalization
head development
nucleic acid phosphodiester bond hydrolysis
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
GPI anchor metabolic process
GPI anchor biosynthetic process
transport
intracellular protein transport
sensory perception of sound
embryonic pattern specification
anterior/posterior axis specification
protein transport
myo-inositol transport
forebrain regionalization
head development
nucleic acid phosphodiester bond hydrolysis
nuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023557 | NM_001163314.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766821 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316096 | AK039452.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165487 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37283 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102551 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157233 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023557 | NM_001163314.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04101221 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04101257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102551 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04101208 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157233 | - | - |
| Whole Mouse Genome G4122A | A_51_P211832 | AK051866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P211832 | AK051360.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P573357 | AK051360.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133283 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102551 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04101221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01124768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00720091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00310098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00310101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00805993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00822597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01124765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00805992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00625353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01020407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00755207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00400112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00796767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00755204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00822599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00400110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00054283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00805991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00660399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00878856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00825760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00755209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00314458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00912336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00209976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00682846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00277276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00755202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01124767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20236951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00209951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20237302 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431169_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436317_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354649 | - | - |
| Mouse Genome 430 2.0 Array | 1431169_at | - | - |
| Mouse Genome 430 2.0 Array | 1436317_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436317_at | - | - |
| Mouse Genome 430 2.0 Array | 1431169_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100019001.mm.1 | NM_001163314.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019001.mm.1 | NM_001163314.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019001.mm.1 | TC0100002349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018999.mm.1 | NM_001163314.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018999.mm.1 | NM_001163314.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018999.mm.1 | TC0100002349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100019000.mm.1 | NM_001163314.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019000.mm.1 | NM_001163314.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019000.mm.1 | TC0100002349.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436317_at | - | - |
| Mouse Expression Set 430 | 1431169_at | - | - |
| Mouse Expression Set 430 | 1431169_PM_at | - | - |
| Mouse Expression Set 430 | 1436317_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431169_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436317_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431169_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436317_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 555618 | - | - |
| ABI Mouse Genome Survey Microarray | 676377 | - | - |

Pgap1
Genomic Map