Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 14 - 15 | 1572 | 56 | |
| GenBank mRNA | AF172276.2 | - | 14 - 15 | 1648 | 56 | |
| AK004700.1 | - | 3 - 4 | 368 | 56 | ||
| AK030109.1 | - | 10 - 11 | 1112 | 56 | ||
| AK149434.1 | - | 14 - 15 | 1533 | 56 | ||
| AY347569.1 | - | 14 - 15 | 1449 | 56 | ||
| AY347570.1 | - | 14 - 15 | 1449 | 56 | ||
| BC049853.1 | - | 14 - 15 | 1524 | 56 | ||
| BC138876.1 | - | 14 - 15 | 1551 | 8861898 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map