Assay Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172695.2 | NP_766283.2 | 4 - 5 | 792 | 98 | |
| GenBank mRNA | AK019070.1 | - | 4 - 5 | 798 | 98 | |
| AK075934.1 | - | 4 - 5 | 724 | 98 | ||
| AK081641.1 | - | 4 - 5 | 683 | 98 | ||
| AK162534.1 | - | 3 - 4 | 2983 | 98 | ||
| AK166122.1 | - | 4 - 5 | 697 | 98 | ||
| BC139355.1 | - | 4 - 5 | 791 | 8862377 | 98 | |
| BC139356.1 | - | 4 - 5 | 791 | 8862378 | 98 | |
| BC139773.1 | - | 4 - 5 | 702 | 40111789 | 98 | |
| M57958.1 | - | 654 | 98 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Plaa
Genomic Map