Assay Details
| Gene Symbol: Casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159344.1 | NP_001152816.1 | 11 - 12 | 3034 | 126 | |
| NM_027195.2 | NP_081471.2 | 11 - 12 | 3034 | 126 | ||
| GenBank mRNA | AK173341.1 | - | 6 - 7 | 1824 | 126 | |
| BC075673.1 | - | 11 - 12 | 3036 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casz1
Genomic Map