Assay Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141931.1 | NP_001135403.1 | 6 - 7 | 886 | 117 | |
| NM_001141932.1 | NP_001135404.1 | 6 - 7 | 886 | 117 | ||
| NM_020296.2 | NP_064692.2 | 6 - 7 | 1079 | 117 | ||
| GenBank mRNA | AB026569.1 | - | 6 - 7 | 802 | 117 | |
| AK133054.1 | - | 6 - 7 | 607 | 117 | ||
| AK135280.1 | - | 6 - 7 | 766 | 117 | ||
| AK145897.1 | - | 6 - 7 | 888 | 117 | ||
| AK147109.1 | - | 6 - 7 | 1080 | 117 | ||
| AK161102.1 | - | 6 - 7 | 836 | 117 | ||
| AK164307.1 | - | 6 - 7 | 880 | 117 | ||
| BC016501.1 | - | 6 - 7 | 1067 | 4527742 | 117 | |
| BC056609.1 | - | 3 - 4 | 278 | 117 | ||
| BC057866.1 | - | 6 - 7 | 918 | 5327143 | 117 | |
More Information
Set Membership: |
|

Rbms1
Genomic Map