Assay Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010021.5 | NP_034151.3 | 8 - 9 | 832 | 74 | |
| GenBank mRNA | AK014854.1 | - | 8 - 9 | 812 | 74 | |
| AK033183.1 | - | 8 - 9 | 812 | 74 | ||
| AK160855.1 | - | 8 - 9 | 691 | 74 | ||
| BC099940.1 | - | 8 - 9 | 792 | 30048233 | 74 | |
| U38690.1 | - | 8 - 9 | 947 | 74 | ||
| U46694.1 | - | 8 - 9 | 728 | 74 | ||
| X95724.1 | - | 8 - 9 | 721 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dazl
Genomic Map