Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 6 - 7 | 779 | 65 | |
| GenBank mRNA | AK046622.1 | - | 6 - 7 | 767 | 65 | |
| AK048952.1 | - | 6 - 7 | 756 | 65 | ||
| AK080952.1 | - | 6 - 7 | 780 | 65 | ||
| AK080963.1 | - | 6 - 7 | 774 | 65 | ||
| AK087396.1 | - | 6 - 7 | 774 | 65 | ||
| AK173257.1 | - | 6 - 7 | 738 | 65 | ||
| BC048706.1 | - | 6 - 7 | 764 | 65 | ||
| BC067204.1 | - | 7 - 8 | 817 | 65 | ||
More Information
Set Membership: |
|

Sbf2
Genomic Map