Assay Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008855.2 | NP_032881.1 | 4 - 5 | 615 | 68 | |
| GenBank mRNA | AK051862.1 | - | 4 - 5 | 608 | 68 | |
| BC127083.1 | - | 4 - 5 | 434 | 40111744 | 68 | |
| BY729820.1 | - | 4 - 5 | 615 | 68 | ||
| CJ050078.1 | - | 4 - 5 | 406 | 68 | ||
| X53532.1 | - | 4 - 5 | 443 | 68 | ||
| X59274.1 | - | 4 - 5 | 431 | 68 | ||
More Information
Set Membership: |
|

Prkcb
Genomic Map