Assay Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009739.3 | NP_033869.1 | 1 - 2 | 92 | 91 | |
| GenBank mRNA | AK145479.1 | - | 1 - 2 | 63 | 91 | |
| AK150607.1 | - | 1 - 2 | 84 | 91 | ||
| AK150723.1 | - | 1 - 2 | 84 | 91 | ||
| AK167519.1 | - | 1 - 2 | 84 | 91 | ||
| BC046595.1 | - | 1 - 2 | 62 | 6313106 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145479.1, AK150607.1, AK150723.1, AK167519.1, BC046595.1, NM_009739.3 transcripts.
Set Membership: |
|

Bckdk
Genomic Map