Assay Details
| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008300.3 | NP_032326.3 | 12 - 13 | 1873 | 69 | |
| GenBank mRNA | AK053601.1 | - | 12 - 13 | 1807 | 69 | |
| AK054211.1 | - | 5 - 6 | 595 | 69 | ||
| AK133772.1 | - | 12 - 13 | 1815 | 69 | ||
| AK148883.1 | - | 1 - 2 | 148 | 69 | ||
| AK155305.1 | - | 12 - 13 | 1790 | 69 | ||
| AK166102.1 | - | 12 - 13 | 1856 | 69 | ||
| AK220167.1 | - | 12 - 13 | 1824 | 69 | ||
| BC003770.1 | - | 12 - 13 | 1782 | 3494648 | 69 | |
| BC003950.1 | - | 12 - 13 | 1768 | 69 | ||
| CA316420.1 | - | 1 - 2 | 168 | 69 | ||
| D85904.1 | - | 12 - 13 | 1575 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4
Genomic Map