Assay Details
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 13 - 14 | 1567 | 91 | |
| NM_001281966.1 | NP_001268895.1 | 13 - 14 | 1558 | 91 | ||
| NM_001281968.1 | NP_001268897.1 | 13 - 14 | 1546 | 91 | ||
| NM_010583.3 | NP_034713.2 | 13 - 14 | 1549 | 91 | ||
| GenBank mRNA | AK037554.1 | - | 629 | 91 | ||
| AK083380.1 | - | 13 - 14 | 1521 | 91 | ||
| AK153613.1 | - | 13 - 14 | 1505 | 91 | ||
| BC128374.1 | - | 13 - 14 | 1475 | 40111687 | 91 | |
| BC128375.1 | - | 13 - 14 | 1472 | 40111688 | 91 | |
| D14042.1 | - | 13 - 14 | 1489 | 91 | ||
| L00619.1 | - | 13 - 14 | 1554 | 91 | ||
| L05631.1 | - | 13 - 14 | 1515 | 91 | ||
| L10628.1 | - | 13 - 14 | 1517 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itk
Genomic Map