Assay Details
| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136070.1 | NP_001129542.1 | 10 - 11 | 1319 | 68 | |
| NM_010709.3 | NP_034839.2 | 10 - 11 | 1319 | 68 | ||
| GenBank mRNA | AK030312.1 | - | 10 - 11 | 1330 | 68 | |
| AK034115.1 | - | 10 - 11 | 1291 | 68 | ||
| AK036154.1 | - | 10 - 11 | 1315 | 68 | ||
| AK046539.1 | - | 10 - 11 | 1331 | 68 | ||
| AK082748.1 | - | 10 - 11 | 1334 | 68 | ||
| AK083638.1 | - | 10 - 11 | 1279 | 68 | ||
| AK139052.1 | - | 10 - 11 | 1319 | 68 | ||
| AK161269.1 | - | 10 - 11 | 1302 | 68 | ||
| AK166085.1 | - | 10 - 11 | 1332 | 68 | ||
| AK167980.1 | - | 10 - 11 | 1315 | 68 | ||
| AK168120.1 | - | 10 - 11 | 1285 | 68 | ||
| AK170252.1 | - | 10 - 11 | 1346 | 68 | ||
| AK170448.1 | - | 10 - 11 | 1331 | 68 | ||
| AK171115.1 | - | 10 - 11 | 1297 | 68 | ||
| BC023709.1 | - | 10 - 11 | 1272 | 68 | ||
| BC024501.1 | - | 5 - 6 | 633 | 68 | ||
| BC025036.1 | - | 10 - 11 | 1275 | 4935223 | 68 | |
| U58337.1 | - | 10 - 11 | 1204 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2d
Genomic Map