Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 10 - 11 | 1421 | 67 | |
| NM_011031.2 | NP_035161.2 | 10 - 11 | 1421 | 67 | ||
| GenBank mRNA | AK142689.1 | - | 2 - 3 | 116 | 67 | |
| AK144124.1 | - | 10 - 11 | 1368 | 67 | ||
| AK145535.1 | - | 10 - 11 | 1348 | 67 | ||
| BC018411.1 | - | 10 - 11 | 1333 | 3157481 | 67 | |
| CD563665.1 | - | 3 - 4 | 279 | 67 | ||
| U16163.1 | - | 9 - 10 | 1308 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha2
Genomic Map